Dimedone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 5,5-Dimethyl-1,3-cyclohexanedione, 5,5-Dimethylcyclohexane-1,3-dione
CAS Number:126-81-8
:
Formula:C₈H₁₂O₂
MDL Number:MFCD00001588
Boiling Pt:234
Molecular Weight:140.18 g/mol
Melting Pt:148
Merck Index:13,03268
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:204-804-4
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together