Phenol red
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
CAS Number:143-74-8
:
Formula:C₁₉H₁₄O₅S
MDL Number:MFCD00003552
Boiling Pt:>300 °C
Molecular Weight:354.38 g/mol
Melting Pt:> 300 °C
Merck Index:13,07329
EINECS:205-609-7
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together