VWR®, Petri Dishes, Glass
Supplier: VWR Collection
Certificates
About this item
Made from borosilicate 3.3 glass that can, therefore, be sterilised and/or used for applications that require heat. These reusable glass Petri dishes are a green alternative to Petri dishes made from plastic, ideal for smaller laboratories.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
- Autoclavable and sterilisable