2,3,5-Trifluorobenzyl alcohol
Supplier: FLUOROCHEM
Certificates
About this item
CAS Number:67640-33-9
:
Formula:C₇H₅F₃O
MDL Number:MFCD00083542
Molecular Weight:162.11 g/mol
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together