3',4'-Dimethoxyacetophenone 98%
: 1-(3,4-Dimethoxyphenyl)ethan-1-one, 3,4-Dimethoxyacetophenone, 1-(3,4-Dimethoxyphenyl)ethanone
:1131-62-0
:
:C₁₀H₁₂O₃
:MFCD00008737
:286…288 °C (760 mmHg)
:180.2 g/mol
:48…52 °C
:>112 °C
:214-468-0
:GHS07
:Ambient
- Environmentally Preferable:
- Appearance:Yellow to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:48 to 52 °C
- GC:≥97.5 %
Frequently Bought Together