2',4'-Dichloroacetophenone 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2′,4′-Dichloroacetophenone
CAS Number:2234-16-4
:
Formula:C₈H₆Cl₂O
MDL Number:MFCD00000581
Boiling Pt:247
Melting Pt:29…34 °C
Density:1.32 g/cm³
Molecular Weight:189.04 g/mol
Flash Pt:131 °C
EINECS:218-780-8
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to yellow-brown Solid
- Infrared spectrum:Conforms
- Melting point:29 to 33 °C
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together