1,4-Dimethoxybenzene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: p-Dimethoxybenzene, Hydroquinone dimethyl ether
CAS Number:150-78-7
:
Formula:C₈H₁₀O₂
MDL Number:MFCD00008401
Boiling Pt:212.6
Melting Pt:54…56 °C
Density:0.874 g/cm³ (24.9 °C)
Molecular Weight:138.17 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:205-771-9
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together