Methyl salicylate, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: Wintergreen Oil, Methyl 2-hydroxybenzoate
CAS Number:119-36-8
:
Formula:2-(HO)C₆H₄CO₂CH₃
MDL Number:MFCD00002214
Boiling Pt:432
Melting Pt:100…101 °C
Density:1.184 g/cm³ (20 °C)
Molecular Weight:152.15 g/mol
Flash Pt:99
Merck Index:13,06143
:M-Clarity™ quality level = MQ100
EINECS:204-317-7
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together