Benzocaine (ethyl-4-aminobenzoate) 98%
: Ethyl 4-aminobenzoate, Aethoform
:94-09-7
:
:C₉H₁₁NO₂
:MFCD00007892
:172 °C (17hPa)
:165.19 g/mol
:89…92 °C
:>100 °C
:13,01087
:202-303-5
:GHS07
:Ambient
- Environmentally Preferable:
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:89 to 92 °C
- GC:≥97.5 %
- Titration with HClO4:≥97.5 %
Frequently Bought Together