Order Entry
United Kingdom
ContactUsLinkComponent
2,6-Dichlorobenzamide 97%
2,6-Dichlorobenzamide 97%
Certificates
undefined
2,6-Dichlorobenzamide 97%

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to grey-brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    196 to 201 °C
  • GC:
    ≥96 %

Specification Test Results

Related Information

Frequently Bought Together