3-Nitrobenzaldehyde, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:99-61-6
:
Formula:C₇H₅NO₃
MDL Number:MFCD00007249
Boiling Pt:232
Melting Pt:58
Density:1.481 g/cm³ (0 °C)
Molecular Weight:151.12 g/mol
Merck Index:12,06684
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:202-772-6
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together