Order Entry
United Kingdom
ContactUsLinkComponent
N-Methyl-2-pyrrolidone (NMP), extra dry 99.5%, AcroSeal®
N-Methyl-2-pyrrolidone (NMP), extra dry 99.5%, AcroSeal®
Certificates
undefined
N-Methyl-2-pyrrolidone (NMP), extra dry 99.5%, AcroSeal®
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.005 %
  • GC:
    ≥99.4 %
  • Color scale:
    ≤20 APHA

Specification Test Results

Frequently Bought Together