cis-2-Butene-1,4-diol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: (Z)-But-2-ene-1,4-diol, (Z)-2-Butene-1,4-diol
CAS Number:6117-80-2
:
Formula:C₄H₈O₂
MDL Number:MFCD00002924
Boiling Pt:235
Density:1.07 g/cm³ (20 °C)
Molecular Weight:88.11 g/mol
Merck Index:12,02390
:M-Clarity™ quality level = MQ100
EINECS:228-085-1
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together