(2,4-Dimethoxyphenyl)acetic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2,4-Dimethoxyphenylacetic Acid
CAS Number:6496-89-5
:
Formula:(C₆H₃(OCH₃)₂)CH₂COOH
MDL Number:MFCD00183328
Molecular Weight:196.2 g/mol
Melting Pt:108…112 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together