L(-)-Glutathione (reduced form) 98% for analysis, reduced
: L-Glutathione Reduced, GSH, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, Glutathione Reduced
:70-18-8
:
:C₁₀H₁₇N₃O₆S
:MFCD00065939
:307.33 g/mol
:182…192 °C (dec.)
:13,04488
:Refrigerator
:200-725-4
- Environmentally Preferable:
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:182 to 192 °C (Dec)
- Titration Iodimetric:≥97.5 %
- Loss on drying:≤0.5 % (105 °C, 3 hrs)
- Sulfated ash:≤0.1 % (800 °C)
- Specific optical rotation:-15.5° to -17.5° (20 °C, 589 nm) (c=4, H2O)
- Arsenic (As):≤2 ppm
- Iron (Fe):≤5 ppm
- Heavy metals (as Pb):≤10 ppm
Frequently Bought Together