Pivalaldehyde (trimethylacetaldehyde), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Pivalaldehyde, Pivaldehyde, Trimethylacetaldehyde, 2,2-Dimethylpropionaldehyde, 2,2-Dimethylpropanal
CAS Number:630-19-3
:
Formula:C₅H₁₀O
MDL Number:MFCD00006962
Boiling Pt:74
Melting Pt:6
Density:0.771 g/cm³ (20 °C)
Molecular Weight:86.13 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Refrigerator
EINECS:211-134-6
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together