Dithiothreitol (DTT, Cleland's reagent), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: cis-2,3-Dihydroxy-1,4-butanedithiol, (R*,R*)-1,4-Dimercapto-2,3-butandiol, DTT, DL-1,4-Dithiothreitol
CAS Number:3483-12-3
:27565-41-9
Formula:C₄H₁₀O₂S₂
MDL Number:MFCD00004877
Boiling Pt:∼125 °C (1013 hPa)
Molecular Weight:154.25 g/mol
Melting Pt:42…44 °C
Flash Pt:>110 °C
:M-Clarity™ quality level = MQ200
EINECS:222-468-7
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together