1,2-Dimethoxyethane for synthesis, Sigma-Aldrich®
: Merck
: Dimethyl glycol, Ethylene glycol dimethyl ether, DME
:110-71-4
:
:C₄H₁₀O₂
:MFCD00008502
:85
:–69 °C
:1.113 g/cm³ (20 °C)
:90.12 g/mol
:–6 °C
:203-794-9
:2252
:3 II
:13,03251
:M-Clarity™ quality level = MQ200
:GHS02
:GHS08
:GHS07
:Ambient
- Environmentally Preferable:
- Solubility:(20 °C) soluble
- Melting Point:-69 °C
- Molar Mass:90.12 g/mol
- Boiling Point:84 - 86 °C (1013 hPa)
- Vapor Pressure:67 hPa (20 °C)
- Flash Point:-6 °C
- Explosion Limit:1.6 - 10.4 %(V)
- Density:0.87 g/cm3 (20 °C)
- pH:(H2O) neutral
- Ignition Point:200 °C
- Viscosity kinematic:0.455 mm2/s (25 °C)
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
Frequently Bought Together