Diethyl ether ≥99.9% inhibitor-free for HPLC, Sigma-Aldrich®
Supplier: Novartis
New Product
Certificates
About this item
Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
CAS Number:60-29-7
:
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646
Boiling Pt:35
Melting Pt:–116 °C (lit.)
Density:0,706 g/ml (25 °C) (lit.)
Molecular Weight:74.12 g/mol
Flash Pt:–40 °C
EINECS:200-467-2
UN:1155
ADR:3 I
Merck Index:12,03852
:M-Clarity™ quality level = MQ200
REACH:01-2119535785-29
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance (colour):Colourless
- Appearance (form):Liquid
- Purity (GC area %):≥99.90%
- Water (coulometr.):≤0.03%
- Residue (evaporation):≤0.0003%
- Infrared spectrum:Conforms to structure
- UV-absorbance:400 nm ≤0.01
- Peroxides:≤1 ppm
- Expiration date period:One year
Specification Test Results
Frequently Bought Together