Order Entry
United Kingdom
ContactUsLinkComponent
 
N,N,N'-Trimethylethylenediamine 97%
 
undefined
N,N,N'-Trimethylethylenediamine 97%

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4180 to 1.4210 (20°C, 589 nm)
  • GC:
    ≥96.0 %

 

 

Frequently Bought Together