Triethanolamine (trolamine) 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Tris-(2-hydroxyethyl)amine, 2,2',2''-Nitrilotriethanol
CAS Number:102-71-6
:
Formula:C₆H₁₅NO₃
MDL Number:MFCD00002855
Boiling Pt:360 °C (760 mmHg)
Melting Pt:21 °C
Density:1.1245 g/cm³
Molecular Weight:149.19 g/mol
Flash Pt:190 °C
Merck Index:13,09739
Storage Temperature:Ambient
EINECS:203-049-8
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.4830 to 1.4860 (20°C, 589 nm)
- GC:≥96.0 %
- Color scale:≤50 APHA
Specification Test Results
Frequently Bought Together