1,1,1,5,5,5-Hexafluoroacetylacetone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Hexafluoroacetylacetone, 1,1,1,5,5,5-Hexafluoropentane-2,4-dione, 1,1,1,5,5,5-Hexafluoro-2,4-pentanedione, HFAA
CAS Number:1522-22-1
:
Formula:C₅H₂F₆O₂
MDL Number:MFCD00000426
Boiling Pt:70
Density:1.47 g/cm³ (20 °C)
Molecular Weight:208.06 g/mol
Flash Pt:32
Storage Temperature:Ambient
EINECS:216-191-0
ADR:8 II
:M-Clarity™ quality level = MQ100
UN:2920
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together