Triammonium citrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 2-Hydroxy-1,2,3-propanetricarboxylic acid triammonium salt, Ammonium citrate tribasic, Citric acid triammonium salt, Triammonium 2-hydroxypropane-1,2,3-tricarboxylate
CAS Number:3458-72-8
:
Formula:C₆H₁₇N₃O₇
MDL Number:MFCD00036406
Density:1 g/cm³ (25 °C)
Molecular Weight:243.22 g/mol
Melting Pt:185
:M-Clarity™ quality level = MQ200
EINECS:222-394-5
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together