Ammonium acetate 7.5 M, clear solution
Supplier: MP Biomedicals
Certificates
About this item
Synonyms: Acetic acid ammonium salt
CAS Number:631-61-8
:
Formula:H₃CCOONH₄
MDL Number:MFCD00013066
Density:1.17 (lit.)
Molecular Weight:77.08 g/mol
Melting Pt:114
Flash Pt:136
Merck Index:13,00495
EINECS:211-162-9
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Identity Test:Passes
- Concentration:7.5 M aq (578.25 g/L)
- Sterility:Sterile
Specification Test Results
Frequently Bought Together