Ethyl-3-mercaptopropionate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Ethyl 3-mercaptopropanoate, 3-Mercaptopropionic acid ethyl ester
CAS Number:5466-06-8
:
Formula:C₅H₁₀O₂S
MDL Number:MFCD00004896
Boiling Pt:75…76 °C (10 mmHg)
Density:1.059 g/cm³
Molecular Weight:134.2 g/mol
Flash Pt:72 °C
Storage Temperature:Refrigerator
EINECS:226-771-5
UN:3334
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4560 to 1.4580 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together