Methyl pyruvate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Pyruvic acid ethyl mester, 2-Ketopropionic acid methyl ester, 2-Oxopropionic acid methyl ester, Methyl 2-oxopropanoate, α-Ketopropionic acid methyl ester
CAS Number:600-22-6
:
Formula:C₄H₆O₃
MDL Number:MFCD00008754
Boiling Pt:137
Density:1.154 g/cm³ (0 °C)
Molecular Weight:102.09 g/mol
Storage Temperature:Refrigerator
EINECS:209-987-4
UN:3272
ADR:3 III
:M-Clarity™ quality level = MQ100
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together