11-Bromoundecanoic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 11-Bromohendecanoic acid, 11-Bromoundecylic acid
CAS Number:2834-05-1
:
Formula:BrH₂C(CH₂)₉COOH
MDL Number:MFCD00002732
Boiling Pt:173…174 °C (2 mmHg)
Molecular Weight:265.19 g/mol
Melting Pt:48…51 °C
Flash Pt:113 °C
EINECS:220-602-9
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to beige Crystalline chunks
- Infrared spectrum:Conforms
- GC:≥97.5 %
- Titration Argentometric:≥94 % (ex Cl)
Specification Test Results
Frequently Bought Together