1,6-Heptadiyne, Sigma-Aldrich®
SIAL407437-1G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
:
:92.14 g/mol
:108
:–85 °C
:0.8164 g/cm³ (17 °C)
:Refrigerator
:219-253-5
:3295
:2396-63-6
:C₇H₈
:MFCD00014925
:3 II
:M-Clarity™ quality level = MQ100