D-(+)-Biotin ≥99%, Reagent Grade
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
CAS Number:58-85-5
:
Formula:C₁₀H₁₆N₂O₃S
MDL Number:MFCD00005541
Density:1.41 g/cm³ (20 °C)
Molecular Weight:244.31 g/mol
Melting Pt:232…233 °C
Merck Index:13,01231
Storage Temperature:Refrigerator
EINECS:200-399-3
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Heavy Metals (as Pb):<0.001%
- Purity (RG):≥99%
- Specific Rotation:89 – 93°
Specification Test Results
Related Information
Frequently Bought Together