L(-)-Tyrosine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: L-Tyrosine, (S)-(-)-Tyrosine, Tyrosine, H-Tyr-OH
CAS Number:60-18-4
:
Formula:C₉H₁₁NO₃
MDL Number:MFCD00002606
Boiling Pt:351
Melting Pt:285
Density:1.41 g/cm³ (20 °C)
Molecular Weight:181.19 g/mol
Merck Index:13,09907
Storage Temperature:Ambient
EINECS:200-460-4
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together