L(+)-Glutamic acid, EMPROVE® ESSENTIAL Ph. Eur., JP suitable for use as excipient, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
Synonyms: (S)-(+)-2-Aminopentanedioic acid, L(+)-1-Aminopropane-1,3-dicarboxylic acid, (S)-(+)-α-Glutamic acid, H-Glu-OH, L-Glutamic acid, L(+)-2-Aminopentanedioic acid, (S)-(+)-Glutamic acid, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid, L(+)-α-Glutamic acid
CAS Number:56-86-0
:
Formula:HO₂CCH₂CH₂CH(NH₂)CO₂H
MDL Number:MFCD00002634
Boiling Pt:325
Melting Pt:160
Density:1.538 g/cm³ (20 °C)
Molecular Weight:147.13 g/mol
Merck Index:13,04482
Storage Temperature:Ambient
EINECS:200-293-7
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together