L(+)-Sodium lactate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Sodium (S)-2-hydroxypropanoate, (S)-(+)-2-Hydroxypropanoic acid sodium salt, (S)-(+)-2-Hydroxypropionic acid sodium salt, Sodium-(S)-lactate, (S)-(+)-Lactic acid sodium salt, L(+)-Lactic acid sodium salt
CAS Number:867-56-1
:
Formula:C₃H₅NaO₃
MDL Number:MFCD00066576
Molecular Weight:112.06 g/mol
:M-Clarity™ quality level = MQ200
EINECS:212-762-3
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together