VWR®, Blocking buffer, non-fat powdered milk
M232-39.8G-5PK
:
- Pk:199 g
- Description:Blocking buffer, PBS with non fat milk, powder blend
- Environmentally Preferable:
Each pack of powdered blocking buffer makes one liter of 1X solution.
- Easy to use
- Optimise assay performance using high quality blocking buffers