Iron(III) nitrate nonahydrate ≥98%, TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Ferric nitrate nonahydrate, Iron trinitrate nonahydrate, Iron(III) nitrate hydrate, Ferric Nitrate, 9-Hydrate
CAS Number:7782-61-8
:
Formula:Fe(NO₃)₃·9H₂O
MDL Number:MFCD00149708
Boiling Pt:125
Melting Pt:47
Density:1.68 g/cm³ (20 °C)
Molecular Weight:404 g/mol
Storage Temperature:Ambient
EINECS:233-899-5
ADR:5.1 III
Merck Index:13,04057
UN:1466
:GHS03
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:Min. 98 %
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 5L Plastic bucket UN Z (ZPACK13247005)
Packaging Information
spec_VWRC24174.296 spec_VWRC24174.296 spec_VWRC24174.365
Product specification
Frequently Bought Together