Manganese(II) nitrate hydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Manganous nitrate hydrate
CAS Number:15710-66-4
:
Formula:Mn(NO₃)₂·H₂O
MDL Number:MFCD00149788
Molecular Weight:196.96 g/mol
ADR:5.1 III
:M-Clarity™ quality level = MQ100
EINECS:233-828-8
UN:2724
:GHS03
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together