Chromium(III) potassium sulphate dodecahydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Chromium potassium sulphate dodecahydrate, Chromium (III) potassium bis(sulphate) dodecahydrate
CAS Number:7788-99-0
:
Formula:CrK(SO₄)₂·12H₂O
MDL Number:MFCD00149917
Density:1.83
Molecular Weight:499.4 g/mol
Melting Pt:89
Merck Index:13,02250
EINECS:233-401-6
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together