Order Entry
United Kingdom
ContactUsLinkComponent
 
Disodium hydrogen phosphate heptahydrate 98.0-102.0% ACS, Fluka™
Catalog # SIALS9390-100G
Supplier:  Merck
CAS Number:  
Disodium hydrogen phosphate heptahydrate 98.0-102.0% ACS, Fluka™
Catalog # SIALS9390-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle, wide mouth
  • Pk:
    100 g
  • Seq:
    3401
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Sodium phosphate dibasic heptahydrate, Sodium hydrogen phosphate heptahydrate, Sodium monohydrogen phosphate heptahydrate
 :
 :268.07 g/mol
Melting Pt:48
Density:1.7 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-448-7
CAS Number:7782-85-6
Formula:HNa₂O₄P·7H₂O
MDL Number:MFCD00149180