Dipotassium hydrogen orthophosphate trihydrate
Supplier: Roth Carl
Certificates
About this item
Synonyms: Dipotassium hydrogen phosphate trihydrate, Potassium hydrogen phosphate trihydrate, di-Potassium hydrogen phosphate trihydrate, Potassium phosphate dibasic trihydrate, di-Potassium hydrogen orthophosphate trihydrate
CAS Number:16788-57-1
:7778-77-0
Formula:K₂HPO₄·3H₂O
MDL Number:MFCD00149926
Boiling Pt:∼340 °C (1013 hPa)
Density:2.504 g/cm³ (20 °C)
Molecular Weight:228.22 g/mol
Storage Temperature:Ambient
EINECS:231-834-5
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together