EDTA disodium salt dihydrate BP, Ph. Eur., ACS, JP, USP
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Ethylenediaminetetraacetic acid, disodium salt, dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium dihydrate, Titriplex III, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Sequestrene Na₂, Edetate disodium dihydrate, Edetic acid disodium salt dihydrate
CAS Number:6381-92-6
:
Formula:C₁₀H₁₄N₂Na₂O₈·2H₂O
MDL Number:MFCD00150037
Molecular Weight:372.24 g/mol
Melting Pt:255
Merck Index:13,03543
Storage Temperature:Ambient
EINECS:205-358-3
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance::99.0 -- 101.0 %
- White or almost white crystalline powder:Passes test
- Heavy metals (as Pb):Max. 10 ppm
- Residual solvents:Passes test
- Residual metal catalysts (skip lot): .:Passes test Ph.Eur./BP
- As (Arsenic):Max. 2 ppm
- Cd (Cadmium):Max. 1 ppm
- Co (Cobalt):Max. 1 ppm
- Hg (Mercury):Max. 3 ppm
- Ni (Nickel):Max. 1 ppm
- Pb (Lead):Max. 3 ppm
- V (Vanadium):Max. 6 ppm
- Conforms to Ph.Eur./USP/BP/JP/ACS:Passes test
- Ph.Eur./BP specifications::Passes test Ph.Eur./BP
- Assay [Ph.Eur./BP]:98.5 --> 101.0 %
- Identification A:Passes test Ph.Eur./BP
- Identification B:Passes test Ph.Eur./BP
- Identification D:Passes test Ph.Eur./BP
- Appearance of solution:Passes test Ph.Eur./BP
- pH (5 %) [Ph.Eur./BP]:4.0 --> 5.5
- Impurity A (*) [Ph.Eur./BP]:Max. 0.1 %
- Fe (Iron) [Ph.Eur./BP]:Max. 80 ppm
- USP specifications::Passes test
- Assay (calculated on dried basis) [USP]:99.0 --> 101.0 %
- Identification C:Passes test USP
- Ca (Calcium):Passes test USP
- Limit of nitrilotriacetic acid [USP]:Max. 0.1 %
- pH (25°C; 50 mg/ml) [USP]:4.0 --> 6.0
- Loss on drying (150°C; 6 h) [USP]:8.7 --> 11.4 %
- Assay [JP]:Min. 99.0 %
- Identification 1:Passes test JP
- Identification 2:Passes test JP
- Identification 3:Passes test JP
- pH (1 %) [JP]:4.3 --> 4.7
- Clarity and colour of solution:Passes test JP
- CN (Cyanide):Passes test JP
- Residue on ignition [JP]:37.0 --> 39.0 %
- Assay [ACS]:99.0 --> 101.0 %
- pH (25°C; 5 %) [ACS]:4.0 --> 6.0
- Insoluble matter [ACS]:Max. 0.005 %
- Nitrilotriacetic acid [ACS]:Max. 0.1 %
- Heavy metals (as Pb) [ACS]:Max. 0.005 %
- Fe (Iron) [ACS]:Max. 0.01 %
Specification Test Results
Related Information
Certificates
Search by Lot Number2,5L Plastic bottle for solids DIN 80 (ZPACK13252500) 20 L Plastic bucket for solids UN Z (ZPACK13247020)
Packaging Information
20309_EDTA DISODIUM SALT 2H2O PH.EUR_ALLERGEN_REV0.pdf Allergen_20309_EDTA DISODIUM SALT 2H2O PH.EUR
ALLERGEN
20309_EDTA DISODIUM SALT 2H2O PH.EUR_BSE_REV1.pdf
BSE/TSE Certificates
20309_EDTA DISODIUM SALT 2H2O PH.EUR_DIOXIN_REV0.pdf 20309_EDTA DISODIUM SALT 2H2O PH.EUR_LATEX_REV0.pdf EDTA Disodium Salt Dihydrate Ph. Eur., USP, BP, JP, ACS
Additional Product Details
27237657.pdf spec_VWRC20309.296 spec_VWRC20309.296 spec_VWRC20309.365 spec_VWRC20309.460
Product specification
Dioxin_20309_EDTA DISODIUM SALT 2H2O PH.EUR.pdf Flowchart_20309_EDTA DISODIUM SALT 2H2O PH.EUR.pdf Latex_20309_EDTA DISODIUM SALT 2H2O PH.EUR.pdf Residual Metal Catalyst_20309_EDTA DISODIUM SALT 2H2O PH.EUR.pdf Solvents_20309_EDTA DISODIUM SALT 2H2O PH.EUR.pdf
Datasheet
VWR Chemicals for Biopharma and Pharma Production
Product Literature
Frequently Bought Together