Order Entry
United Kingdom
ContactUsLinkComponent
Zinc acetate dihydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Zinc acetate dihydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Zinc acetate dihydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Insolubility in water:
    Max. 50 ppm
  • Not precipitated by (NH4)2S (as SO4):
    Max. 0.25 %
  • Reducing substances (as HCOOH):
    Max. 0.015 %
  • Cl (Chloride):
    Max. 5 ppm
  • NH4 (Ammonium):
    Max. 10 ppm
  • NO3 (Nitrate):
    Max. 10 ppm
  • SO4 (Sulphate):
    Max. 20 ppm
  • As (Arsenic):
    Max. 2 ppm
  • Cu (Copper):
    Max. 10 ppm
  • Fe (Iron):
    Max. 5 ppm
  • Pb (Lead):
    Max. 20 ppm

Specification Test Results

Frequently Bought Together