Order Entry
United Kingdom
ContactUsLinkComponent
 
Phenylhydrazinium chloride Reag. Ph. Eur. analytical reagent
 :  Merck
 
undefined
Phenylhydrazinium chloride Reag. Ph. Eur. analytical reagent
 :  Merck
 

 

  • Environmentally Preferable:

 

 

  • Solubility:
    50 g/l (20 °C)
  • Melting Point:
    242 - 246 °C (decomposition)
  • Molar Mass:
    144.6 g/mol
  • Bulk Density:
    330 kg/m3
  • pH:
    2.6 - 2.9 (50 g/l, H2O)

 

Frequently Bought Together