Order Entry
United Kingdom
ContactUsLinkComponent
 
Hydrazine hydrate 100% (∼64% w/w N₂H₄)
 
undefined
Hydrazine hydrate 100% (∼64% w/w N₂H₄)
 

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Clear colorless Liquid
  • Refractive index:
    1.4285 to 1.4315 (20°C, 589 nm)
  • Density:
    (20°C) 1.030 to 1.033 g/ml
  • Titration with HCl:
    63.6 to 64.6 % (N2H4)

 

 

Frequently Bought Together