Oxalic acid 10% in aqueous solution 10% (w/v) in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Ethanedioic acid
CAS Number:144-62-7
:
Formula:C₂H₂O₄
MDL Number:MFCD00002573
Boiling Pt:365
Molecular Weight:90.04 g/mol
ADR:8 III
Merck Index:13,06980
EINECS:205-634-3
UN:3265
:GHS05
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together