2-Amino-6-methylheptane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 6-Methyl-2-heptylamine, Octodrine
CAS Number:543-82-8
:
Formula:C₈H₁₉N
MDL Number:MFCD00008092
Boiling Pt:154…156 °C (1013 hPa)
Density:0.767 g/cm³ (20 °C)
Molecular Weight:129.25 g/mol
Storage Temperature:Ambient
ADR:8 II
Merck Index:12,06854
:M-Clarity™ quality level = MQ100
EINECS:208-851-1
UN:2734
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together