Polyvinylpyrrolidone, M.W. ∼ 8.000 (K16-18)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP
CAS Number:9003-39-8
:
Formula:(C₆H₉NO)n
MDL Number:MFCD01076626
Melting Pt:130 °C
Merck Index:14,7697
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together