Polysorbate 85
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Sorbitan trioleate, ethoxylated, Polyoxyethylenesorbitan Trioleate
CAS Number:9005-70-3
:
MDL Number:MFCD01779641
Density:1.02 g/cm³
Flash Pt:>200 °C
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Yellow to amber Viscous liquid
- Infrared spectrum:Conforms
- Water:≤3 %
- Acid value:≤2 mg KOH/g
- Hydroxyl value:38 to 52 mg KOH/g
- Saponification value:80 to 95 mg KOH/g
Specification Test Results
Frequently Bought Together