Tween® 20 (Polysorbate) for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Polyoxyethylene sorbitan monolaurate
CAS Number:9005-64-5
:
Formula:C₅₈H₁₁₄O₂₆
MDL Number:MFCD00165986
Density:1.1 g/cm³ (20 °C)
Molecular Weight:1227.53 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:500-018-3
Limited Time Sale
Select items below are currently on sale
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(25 °C) soluble
- Molar Mass:1228 g/mol
- Boiling Point:100 °C
- Vapor Pressure:- 1.4 hPa (20 °C)
- Flash Point:149 °C
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC8.22184.0500_TDS_EN MERC8.22184.1000_TDS_EN ProductPage_822184
Product specification
Merck Individual products and tailor-made chemicals for your technical applications
Product Literature
msds_822184
SDS
Frequently Bought Together