TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) analytical reagent, Millipore®
Supplier: Merck
Certificates
About this item
Synonyms: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol
CAS Number:9036-19-5
:9002-93-1
MDL Number:MFCD00132505
Boiling Pt:>200 °C (1013 hPa)
Melting Pt:6
Density:1.07 g/cm³ (20 °C)
Flash Pt:251 °C (closed cup)
Storage Temperature:Ambient
ADR:9 III
Merck Index:13,06793
:M-Clarity™ quality level = MQ200
UN:3082
:GHS05
:GHS07
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Identity (IR spectrum):passes test
- pH value (10 g/l, H₂O, 20 °C):5.0 - 8.0
- Density (d 20/4):1.064 - 1.067
- Cloud point:63 - 69 °C
- Ethylene oxide (GC):≤30 ppm
- Dioxane (GC):≤20 ppm
- Sulphated ash (600 °C):≤0.2%
- Water (acc. to Karl Fischer):≤0.2%
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.08603.1000_TDS_EN ProductPage_108603
Product specification
msds_108603
SDS
Frequently Bought Together