Silicone fluid, 50 cSt DC 200
Catalog # SERA35134.01
Supplier: Serva
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
Specifications
About this item
Synonyms: Dimethylpolysiloxane, PDMS, Dimethyl siloxane polymer (methyl silicones), Silicone oil, Dimethicone, Polydimethylsiloxane
:
Storage Temperature:Ambient
CAS Number:63148-62-9
:9016-00-6
MDL Number:MFCD00084411
Density:0.963 g/cm³ (25 °C)