Canada balsam
Catalog # SERA26896.02
Supplier: Serva
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 ml
- Seq:2440
- Environmentally Preferable:
Specifications
About this item
:
Density:0.915 g/cm³ (20 °C)
Flash Pt:62
Storage Temperature:Ambient
EINECS:232-362-2
CAS Number:8007-47-4
MDL Number:MFCD00132800
Merck Index:13,00948